Learn More
Vernier™ Digital Protoboard Adapter
SDPSCI_ED
Catalog No. S16106ND Shop All Vernier Software Products
Change view
Click to view available options
Warranty:
5 Year Limited
Catalog No. Warranty
S16106ND 5 Year Limited
1 options
Catalog No. S16106ND Supplier Vernier™ Supplier No. BTDELV
Add to Cart
Add to Cart
Includes
Digital Protoboard Adapter
Requires
For use with standard prototyping boards or National Instruments ELVIS.
This connector makes it easy to wire the signal from a Vernier digital sensor into a non-Vernier interface.
  • The adapter fits into a standard prototyping board or National Instruments' ELVIS prototyping board, providing easy access to the sensor lines
Warranty and Services
5-year limited warranty
Specifications
Warranty 5 Year Limited
For Use With (Application) Educational use only
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov