Learn More
Vernier™ Digital Sensor Cable
SDPSCI_ED
Catalog No. S16197ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
CBR 2, Go!Motion, Motion Detector
Catalog No. For Use With (Equipment)
S16197ND CBR 2, Go!Motion, Motion Detector
1 options
Catalog No. S16197ND Supplier Vernier™ Supplier No. MDCBTD
Add to Cart
Add to Cart
Includes
Digital Sensor Cable
This cable connects Vernier digital sensors to the digital (DIG) port on a Vernier interface.
  • Compatible Sensors and Accessories
    • Motion Detector (included with sensor)
    • Drop Counter (included with sensor)
    • Go!Motion
Warranty and Services
5-year limited warranty
Specifications
Interface DIG Port
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) CBR 2, Go!Motion, Motion Detector