Learn More
Market Lab DIGITAL THERMOMETER BARE WIRE
Supplier:  Market Lab 52431
Encompass_Preferred
Digital Workstation Thermometer with Bare Wire Probe.Measuring range: -83°-392°F; accuracy: ±0.9°F.Prove wire is 1mm in diameter for ease of use.
Catalog No. NC2000616
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.