Learn More
United Scientific™ Digital Timer with Photogates
SDPSCI_ED
Catalog No. S05928 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Time Range:
0.00 min. to 9999 sec.
Catalog No. Time Range
S05928 0.00 min. to 9999 sec.
1 options
This item is not returnable. View return policy
Catalog No. S05928 Supplier United Scientific™ Supplier No. DTPHG1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microprocessor-controlled four-digit Digital Timer with Photogates.
  • Designed to be used with air tracks, free-fall apparatus and other equipment.
  • Offers eight programmed functions to record counts, time intervals, periods of oscillation, accelerations, acceleration due to gravity and velocities in collisions.
  • Comes with two photogates.
Specifications
Product Type Digital Timer
Time Range 0.00 min. to 9999 sec.
Alarms Audible/Visible
Display LED
Weight (English) 5.2 in.
Weight (Metric) 2.35 kg
Includes Unit Indicators, function indicator, on/off switch, free fall switch, stop button, data display, function selector, clear/zero button, 2 photogates with support rods and screws
For Use With (Application) Measuring the acceleration of gravity
For Use With (Equipment) Air tracks, free-fall apparatus and other equipment