Learn More
Laxco™ Digital Upgrade Kit
GREENER_CHOICESDP
This Digital Upgrade Kit provides image capture capabilities to microscopes. Options for SLi3 PRO Series and SLi6 PRO Series Microscopes.
$7665.00 - $7770.00
Specifications
Includes Camera, Image Capture Software, Mount Adaptor
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Camera Type CMOS
Display Type Computer
Product Type Microscope Camera
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
SLI3P-DU-KIT3
GREENER_CHOICESDP
View Documents
Laxco™
SLI3PDUKIT3
SLi3 PRO Series Microscopes
Each for $7,665.00
Only null left
Add to Cart
SLI6P-DU-KIT2
GREENER_CHOICESDP
View Documents
Laxco™
SLI6PDUKIT2
SLi6 PRO Series Microscopes
Each for $7,770.00
Only null left
Add to Cart
Description
Description
  • SeBaCamCool 1.7 C Cooled Color 1.7 Megapixel CMOS Camera
  • SeBalit Software-Fluorescent Image Capture Software
  • Mount adaptor included
  • Options available for use with SLi3 PRO Series Microscopes or SLi6 PRO Series Microscopes
Specifications
Specifications
Camera, Image Capture Software, Mount Adaptor
CMOS
Microscope Camera
5 years optical and mechanical, 1 year electrical from date of purchase
Computer
1.7 MP
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.