Learn More
Ansell™ Digitcare™ SafeGlo™ SG12 Medical Exam Gloves
Catalog No. 19500989 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
19-500-989 X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-500-989 Supplier Ansell™ Supplier No. SG124
This item is not returnable. View return policy
Ansell™ Digitcare™ SafeGlo™ SG12 Medical Exam Gloves designed for emergency workers in high-risk applications
  • Advanced non-latex nitrile polymer
  • Polymer coating for easier donning
  • AQL of 0.65 exceeds industry standards
  • SafeGlo™ orange color for high visibility and improved first responder safety
  • ExactFit™ glove formers for reduced hand fatigue and improved dexterity
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D6319
Color Orange
Disposable Yes
Latex Free Yes
Material Nitrile Polymer
Size X-Large
Packaging Type Box
Quantity 100/Pk.
Length (English) 12 in.