Learn More
Description
Sheep anti digoxigenin antibody recognizes digoxigenin, a steroid found in plants that is often attached to oligonucleotides and used as a molecular probe to detect mRNA production.
Specifications
Specifications
| Antigen | Digoxigenin |
| Applications | ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene Alias | C23H34O5; DIG; Digitalis |
| Host Species | Sheep |
| Immunogen | Digoxin-3-CM conjugated to KLH. |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
