Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Digoxin Monoclonal Antibody (38G12)
GREENER_CHOICE

Catalog No. PIMA514746 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514746 Supplier Invitrogen™ Supplier No. MA514746
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA5-14746 targets Digoxin in Western Blot, ELISA, IP and RIA applications and shows reactivity with labeled oligonucleotides and chemical samples. Product MA514746 is a smaller package size of MID0302 (formerly sold as a Seradyn product).

Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Digoxin
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 38G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene Alias C41H64O14; DGX; Lanoxin
Host Species Mouse
Immunogen Digoxin-BSA.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.