Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihexylamine, 99+%

Catalog No. AC407595000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407595000 500 g Glass bottle
AC407591000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407595000 Supplier Thermo Scientific Chemicals Supplier No. 407595000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 143-16-8 | C12H27N | 185.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-16-8
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.35
MDL Number MFCD00009521
InChI Key PXSXRABJBXYMFT-UHFFFAOYSA-N
Synonym dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
PubChem CID 8920
IUPAC Name N-hexylhexan-1-amine
SMILES CCCCCCNCCCCCC

Specifications

Melting Point 3°C
Color Colorless
Density 0.8000g/mL
Boiling Point 236°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4320 to 1.434
Linear Formula (CH3(CH2)5)2NH
Quantity 500 g
UN Number 2927
Beilstein 04,I,384
Specific Gravity 0.8
Solubility Information Solubility in water: 0.3g/L(25°C)
Formula Weight 185.35
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dihexylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-588-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.