Learn More
Medchemexpress LLC Dihydroartemisinin (mixture of α and β isomers) | 131175-87-6 | MFCD00274495 | 98.0% | 284.35 g/mol | C15H24O5 | 100 MG
Supplier: Medchemexpress LLC HYW129233100MG
Dihydroartemisinin (mixture of α and β isomers) is an active metabolite of artemisinin used in research as an antimalarial and anticancer agent. It modulates cell death and autophagy pathways and is used in studies of parasite infection, apoptosis, and tumor biology.
- High purity: 98.0%.
- Molecular formula: C15H24O5; molecular weight: 284.35 g/mol.
- Available in research pack sizes including 100 MG.
- Active metabolite of artemisinin with antimalarial activity.
- Induces apoptosis and autophagy in cellular models.
- Powder storage: -20°C (3 years) or 4°C (2 years).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.