Learn More
Medchemexpress LLC Dihydroartemisinin (mixture of α and β isomers) | 131175-87-6 | MFCD28384156 | 98.0% | 284.35 g/mol | C15H24O5 | 50 MG
Supplier: Medchemexpress LLC HYW12923350MG
Dihydroartemisinin (mixture of α and β isomers) is an antimalarial and anticancer research compound and the active metabolite of artemisinin derivatives. Supplied as a solid for laboratory use, it is intended for research and analytical applications where a high-purity reference or test material is required. Typical identifiers include CAS 131175-87-6, molecular formula C15H24O5, and molecular weight 284.35 g/mol.
- High purity 98.0% suitable for research
- Mixture of α and β isomers for representative biological activity
- Solid form for storage and compound handling
- Common analytical identifiers provided for traceability
- Suitable for antimalarial and anticancer studies