Learn More
Apexbio Technology LLC Artemisinine 63968-64-9 500mg
SDP
Supplier:  Apexbio Technology LLC N1827500
Encompass_Preferred
Artemisinine (CAS 63968-64-9) is a sesquiterpene lactone designed to modulate parasite cell integrity and cancer cell viability Artemisinine exerts its biological activity primarily through interaction with parasite-derived heme groups leading to the formation of reactive oxygen species that disrupt cell integrity and by inducing oxidative stress and apoptotic pathways in tumor cells In in vitro studies Artemisinine demonstrates cytotoxicity toward cancer cells and antimalarial activity with reported IC50 values ranging from 5 to 20 M depending on experimental conditions cell or parasite lines utilized Based on these pharmacological properties Artemisinine holds research potential in antimalarial and antitumor applications
Catalog No. 50-000-90714
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.