Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydroergotamine methanesulfonate

Catalog No. AAJ6384003
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6384003 1 g
AAJ63840MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6384003 Supplier Thermo Scientific Chemicals Supplier No. J6384003
Add to Cart

CAS: 6190-39-2 | C34H41N5O8S | 679.79 g/mol

Agonist at vascular serotonin receptors; partial agonist at alpha-adrenergic and dopamine D2 receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Agonist at vascular serotonin receptors
  • Partial agonist at alpha-adrenergic and dopamine D2 receptors
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6190-39-2
Molecular Formula C34H41N5O8S
Molecular Weight (g/mol) 679.79
MDL Number MFCD00058615
InChI Key ADYPXRFPBQGGAH-UMYZUSPBSA-N
Synonym dihydroergotamine mesylate, dihydroergotamine methanesulfonate salt, dihydroergotoxine, prestwick_746, dihydroergotamine methanesulfonate salt, powder, dihydroergotamine mesilate, european pharmacopoeia ep reference standard, dihydroergotamine for peak identification, european pharmacopoeia ep reference standard, dihydroergotamine mesylate, united states pharmacopeia usp reference standard, 4r,7r-n-1s,2s,4r,7s-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo 7.3.0.0^ 2,6 dodecan-4-yl-6-methyl-6,11-diazatetracyclo 7.6.1.0^ 2,7 .0^ 12,16 hexadeca-1 15 ,9,12 16 ,13-tetraene-4-carboxamide; methanesulfonic acid, 6ar,9r-n-2r,5s,10as,10bs-5-benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8h-oxazolo 3,2-a pyrrolo 2,1-c pyrazin-2-yl-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo 4,3-fg quinoline-9-carboxamide methanesulfonate
PubChem CID 6420006
IUPAC Name (2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
SMILES CS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O

Specifications

Melting Point 230°C to 235°C
Color Pink to White
Quantity 1 g
Merck Index 14,3174
Formula Weight 679.78
Physical Form Powder
Chemical Name or Material Dihydroergotamine methanesulfonate
Hazard Category H312+H332
Hazard Statement GHS H Statement
H312-H332
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
EINECSNumber 228-235-6
RTECSNumber KE7920000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.