Description
Dihydroethidium has been used as a superoxide probe in bovine pulmonary artery endothelial cell (BPAEC). It has been used to check the in situ generation of ROS (reactive oxygen species).
Specifications
Specifications
| Quantity | 10 mg |
| Detection Method | Live Cell Imaging |
| Indicator | ROS |