Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrolinalool 97.0+%, TCI America™
SDP

Catalog No. D159225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D159225ML 25 mL
D1592500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D159225ML Supplier TCI America Supplier No. D159225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18479-51-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00059140
InChI Key JRTBBCBDKSRRCY-UHFFFAOYSA-N
Synonym 3,7-Dimethyl-6-octen-3-ol
PubChem CID 86749
IUPAC Name 3,7-dimethyloct-6-en-3-ol
SMILES CCC(C)(CCC=C(C)C)O

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 200°C
Quantity 25 mL
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dihydrolinalool
EINECSNumber (2)-0258
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.