Learn More
Cayman Chemical Dihydrorhodamine 123, 109244-58-8, 25 mg
SDP
Supplier:  Cayman Chemical 8510025
Encompass_Preferred
A fluorophore that can be used as an indicator of peroxynitrite formation; neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DHR; used to investigate reactive oxygen intermediates produced by endothelial cells, eosinophils, and reactive microglia
Catalog No. NC1005033
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.