Description
Dihydrorhodamine 123 (DHR) has been used for the detection of free intracellular radicals or ROS (reactive oxygen species) in head kidney-derived macrophages. It has also been used in DHR assay in red blood cells and in oxidative burst assay in neutrophils.
Specifications
Specifications
| Quantity | 10 mg |
| Detection Method | Live Cell Imaging |
| Indicator | ROS |