Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dihydroxyacetone Phosphate Assay Kit, Fluorometric, 100 Tests

Catalog No. 502589488
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-9488 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-9488 Supplier Abcam Supplier No. AB197003100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sensitive, quantitative fluorometric assay for measuring dihydroxyacetone phosphate in diverse biological samples.

  • Detection method: Fluorescent
  • Sensitivity: > 0.5 μM
  • Results type: Quantitative
  • Sample types: Plasma, Tissue, Suspension cells, Serum, Other biological fluids, Adherent cells
  • Equipment: Microplate reader
  • Citations: > 3 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Dihydroxyacetone Phosphate
Substrate Properties Chemical Substrate
No. of Reactions 100 Tests
For Use With (Equipment) Microplate Reader
Detection Location In-Solution Detection
Format Kit
Detection Method Fluorescence
Sample Type Plasma, Tissue, Suspension cells, Serum, Other biological fluids, Adherent cells
Quantity 100 Tests
For Use With (Application) Measurement of Dihydroxyacetone phosphate in biofluids and cell or tissue extracts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.