Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutyl Fumarate 98.0+%, TCI America™
SDP

Catalog No. D391525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D391525G 25 g
D3915500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D391525G Supplier TCI America Supplier No. D391525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7283-69-4
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD00152430
InChI Key RSRICHZMFPHXLE-AATRIKPKSA-N
Synonym Fumaric Acid Diisobutyl Ester
PubChem CID 5373437
IUPAC Name 1,4-bis(2-methylpropyl) (2E)-but-2-enedioate
SMILES CC(C)COC(=O)\C=C\C(=O)OCC(C)C

Specifications

Melting Point 8°C
Color Colorless
Boiling Point 122°C
Quantity 25 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutyl Fumarate
EINECSNumber (2)-1099
RTECSNumber LT1565000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.