Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Adipate, +99%, Spectrum™ Chemical
SDP

Catalog No. 18620605 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-605 25 mL
18-620-606 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-605 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D211825ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Synonym: Adipic Acid Diisopropyl Ester, Isopropyl Adipate, Dipropan-2-yl Hexanedioate

  • Used in cosmetic and personal care products as an emollient and fragrance
  • Ungraded products are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use

Chemical Identifiers

CAS 6938-94-9

Specifications

Boiling Point 120°C (248°F)
Quantity 25 mL
Molecular Weight (g/mol) 230.3
Percent Purity 99%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material Diisopropyl Adipate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.