Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Adipate 99.0+%, TCI America™
SDP

Catalog No. A065425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A065425ML 25 mL
A0654500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A065425ML Supplier TCI America Supplier No. A065425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6938-94-9
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.30
MDL Number MFCD00026391
InChI Key ZDQWESQEGGJUCH-UHFFFAOYSA-N
Synonym Adipic Acid Diisopropyl Ester
PubChem CID 23368
ChEBI CHEBI:34711
IUPAC Name 1,6-bis(propan-2-yl) hexanedioate
SMILES CC(C)OC(=O)CCCCC(=O)OC(C)C

Specifications

Melting Point -1°C
Color Yellow
Boiling Point 120°C
Quantity 25 mL
Formula Weight 230.30
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AV1575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.