Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl cyanomethylphosphonate, 97%, Thermo Scientific™

Catalog No. AC224260250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC224260250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC224260250 Supplier Thermo Scientific Chemicals Supplier No. 224260250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

CAS 58264-04-3
Density 1.0300g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C8H16NO3P
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Linear Formula [(CH3)2CHO]P(O)CH2CN
Quantity 25g
MDL Number MFCD00009812
InChI Key MQDIIJHQSUNBKA-UHFFFAOYSA-N
SMILES CC(C)OP(=O)(CC#N)OC(C)C
IUPAC Name bis(propan-2-yl) (cyanomethyl)phosphonate
Molecular Weight (g/mol) 205.19
Formula Weight 205.2
Percent Purity ≥96.0%
Chemical Name or Material Diisopropyl cyanomethylphosphonate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.