Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl ether, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018777 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8777 5 mL
11-102-0398 1 mL
11-101-8344 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8777 Supplier MilliporeSigma Supelco Supplier No. 952515MLF
Add to Cart
Add to Cart

Diisopropyl ether is a secondary ether , which finds applications as an extractant and oxygenate gasoline additive.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 827°F
CAS 108-20-3
Vapor Pressure 120 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.725 g/mL (at 25°C)
Flash Point -29°C
Physical Form Neat
Linear Formula (CH3)2 CHOCH(CH3)2
MDL Number MFCD00008880
Melting Point -85°C
Boiling Point 68°C to 69°C
Synonym Isopropyl ether
Type Analytical Standard
Quantity 5 mL
Formula Weight 102.17
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.