Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl ether, puriss p.a., ≥98.5% (GC), Solstice

Catalog No. 60048428 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-428 2.5 L Glass bottle
60-048-427 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-428 Supplier Solstice Supplier No. 382702.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥98.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -86°C
Density 0.724g/mL at 20°C, 0.725g/mL at 25°C
Boiling Point 67°C to 70°C
Flash Point −29°C (−20.2°F)
Packaging Glass bottle
Refractive Index 1.367
Linear Formula (CH3)2CHOCH(CH3)2
Quantity 2.5 L
UN Number UN1159
Beilstein 1731256
Vapor Density 3.5 (vs air)
Formula Weight 102.17g/mol
Percent Purity ≥98.5% (GC)
Grade ACS/puriss. p.a.
Physical Form Liquid
Chemical Name or Material Diisopropyl ether
Show More Show Less
Hazard Statement P210-P261-P273
DOTInformation class 3 - PG 2 - Diisopropyl ether
RTECSNumber 203-560-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.