Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Hydrazine-1,2-dicarboxylate 98.0+%, TCI America™
SDP

Catalog No. D53651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53651G 1 g
D5365200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53651G Supplier TCI America Supplier No. D53651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19740-72-8
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.23
MDL Number MFCD19443326
InChI Key FBZULTVJWVCJQV-UHFFFAOYSA-N
Synonym Hydrazine-1,2-dicarboxylic Acid Diisopropyl Ester
PubChem CID 88220
IUPAC Name N'-[(propan-2-yloxy)carbonyl](propan-2-yloxy)carbohydrazide
SMILES CC(C)OC(=O)NNC(=O)OC(C)C

Specifications

Melting Point 108°C
Color White
Quantity 1 g
Formula Weight 204.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diisopropyl Hydrazine-1,2-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.