Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Phthalate 98.0+%, TCI America™
SDP

Catalog No. P030125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P030125ML 25 mL
P0301500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P030125ML Supplier TCI America Supplier No. P030125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 605-45-8
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.29
MDL Number MFCD00053717
InChI Key QWDBCIAVABMJPP-UHFFFAOYSA-N
Synonym diisopropyl phthalate, phthalic acid, diisopropyl ester, diisopropyl phthalic acid, unii-bfc138q4px, phthalic acid diisopropyl ester, 1,2-benzenedicarboxylic acid, bis 1-methylethyl ester, bfc138q4px, 1,2-benzenedicarboxylic acid, 1,2-bis 1-methylethyl ester, methylethyl 2-methylethyl oxycarbonyl benzoate, diisopropylphthalate
PubChem CID 11799
ChEBI CHEBI:34712
IUPAC Name 1,2-bis(propan-2-yl) benzene-1,2-dicarboxylate
SMILES CC(C)OC(=O)C1=CC=CC=C1C(=O)OC(C)C

Specifications

Color White
Quantity 25 mL
Formula Weight 250.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Diisopropyl Phthalate
RTECSNumber TI1350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.