Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate, 97%, Thermo Scientific™

Catalog No. ac435700010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435700010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435700010 Supplier Thermo Scientific Chemicals Supplier No. 435700010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 369405-27-6
Molecular Formula C13H29BF4N2
Molecular Weight (g/mol) 300.19
MDL Number MFCD15144860
InChI Key PEKVUVXZBLFOHV-UHFFFAOYSA-N
Synonym n,n,n',n'-tetraisopropylformamidinium tetrafluoroborate, diisopropylamino methylidene diisopropylazanium tetrafluoroborate, bis-diisopropylamino methylium tetrafluoroborate, n-diisopropylamino methylene-n-diisopropylaminium tetrafluoroborate, n-diisopropylamino methylene-n-isopropylpropan-2-aminium tetrafluoroborate, n,n,n inverted exclamation marka,n inverted exclamation marka-tetraisopropylformamidinium tetrafluoroborate
PubChem CID 11077431
IUPAC Name tetrafluoroboranuide; {[bis(propan-2-yl)amino]methylidene}bis(propan-2-yl)azanium
SMILES F[B-](F)(F)F.CC(C)N(C=[N+](C(C)C)C(C)C)C(C)C

Specifications

Melting Point 280.0°C to 284.0°C
Color White to Beige
Physical Form Powder/Solid
Quantity 1 g
Assay Percent Range 97%
Formula Weight 300.19
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.