Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dilauryldimethylammonium Bromide 98.0+%, TCI America™
SDP

Catalog No. D1974100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1974100G 100 g
D197425G 25 g
D1974500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1974100G Supplier TCI America Supplier No. D1974100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3282-73-3
Molecular Formula C26H56BrN
Molecular Weight (g/mol) 462.645
MDL Number MFCD00041969
InChI Key XRWMGCFJVKDVMD-UHFFFAOYSA-M
Synonym didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1
PubChem CID 18669
IUPAC Name didodecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-]

Specifications

Melting Point 161°C
Color White
Quantity 100 g
Formula Weight 462.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dilauryldimethylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.