Learn More
Medchemexpress LLC Diltiazem-d3 hydrochloride | 1217623-80-7 | MFCD21364781 | 98.7% | 454.00 g/mol | C22H24D3ClN2O4S | 5 MG
Supplier: Medchemexpress LLC HY14656S5MG
Diltiazem-d3 hydrochloride is the deuterium-labeled form of diltiazem hydrochloride provided as an isotope-labeled reference standard for analytical and research use. It is the hydrochloride salt with three deuterium atoms (d3) and is intended primarily as an internal standard for quantitative mass spectrometry and pharmacokinetic studies.
- Deuterium-labeled internal standard for MS-based quantitation.
- Hydrochloride salt form, solid suitable for handling and storage.
- High purity suitable for analytical use (≈98.7%).
- Supplied in small milligram packages for reference applications.
- Provides isotopic shift for clear distinction without altering core pharmacology.