Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Diltiazem Hydrochloride USP
SDP

Catalog No. 14186431 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-431 5 g
14-185-617 500 g
14-185-276 25 g
14-185-157 10 g
14-185-380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-186-431 Supplier Medisca Supplier No. 022703
Add to Cart
Add to Cart

Diltiazem Hydrochloride USP is a white or almost white, odorless, crystalline powder or small crystals.

Diltiazem Hydrochloride USP is a white or almost white, odorless, crystalline powder or small crystals.

Specifications

Chemical Name or Material Diltiazem Hydrochloride USP
Packaging Jar 30ml, White, HDPE, Round, 28-400, TRIVENI-DB, CT
CAS 33286-22-5
Type Drug Standard
Molecular Weight (g/mol) 450.98
Quantity 5 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.