Learn More
Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Description
Achieve maximum productivity with the Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories. Expand the flexibility and maintain long-term functionality of the two platforms with accessories that enable the use of different media formats (bottles/bags), enhance the user interface and support proper function of the system.
- Alternative receptacles (bottles instead of bags)
- Alternative media sources (broth bags/bottles)
- Additional/replacement bag stands for ease of preparation
- Bag storage for incubation
- Sample labeling
- Maintenance with replacement parts
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Product Type | Weight |
| Description | Tube weight (5 pieces) |
| For Use With (Equipment) | Diluflux™ Diluter and Homogenizer |
| Packaging Quantity | 1 Ea. |
For research use only.