Learn More
Sigma Aldrich Fine Chemicals Biosciences Plant DNA Dilution Solution | Quantity: 120 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D5688120ML
Encompass
Dilution solution | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MFCD: N/A | 120mLA reagent that dilutes plant DNA extracts and removes PCR inhibitors prior to amplification. Allows direct PCR from plant samples by reducing substances inhibitory to PCR. For use with plant DNA extraction and plant PCR kits to enable plant DNA PCR and amplification.
Catalog No. NC2455512
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.