Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIM-C-pPhOCH3

Catalog No. p-7116133
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-762-5 25 mg
63-765 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-762-5 Supplier R&D Systems Supplier No. 6376/25
This item is not returnable. View return policy

Nur77 agonist

DIM-C-pPhOCH3 is a Nur77 agonist. Induces apoptosis in cancer cell lines in vitro. Enhances long-term potentiation in hippocampal slices and long-term contextual fear memory in an in vivo mouse model.

Specifications

Chemical Name or Material 1,1-Bis(3'-indolyl)-1-(p-methoxyphenyl)methane
CAS 33985-68-1
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C24H20N2O
Formula Weight Observed MW: 352.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.