Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dimaprit Dihydrochloride, Selective H2 Agonist, Solid, 100 mg

Catalog No. 502707195
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7195 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7195 Supplier Abcam Supplier No. AB120855100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective H2 agonist that is systemically active following oral administration.

  • Molecular weight: 234.19 Da
  • Selective H2 agonist with some selectivity for H3
  • Systemically active following oral administration
  • Does not readily cross the blood-brain barrier
  • Physical state: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Target Molecule H2 Receptor, H3 Receptor
Buffer 100% Dimaprit dihydrochloride
Formulation 100% Dimaprit dihydrochloride
Quantity 100 mg
For Use With (Application) H2 receptor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.