Learn More
Creative Biomart Etanercept, Recombinant human TNFR, Fc tagged
Supplier:  Creative Biomart THP0112
Encompass_Preferred
Dimeric fusion protein consisting of the extracellular ligand-binding portion of the human 75 kilodalton (p75) tumor necrosis factor receptor (TNFR) linked to the Fc portion of human IgG1. The Fc component of the product contains the CH2 domain, the CH3 domain and hinge region, but not the CH1 domain of IgG1. The product is produced by recombinant DNA technology in a Chinese hamster ovary (CHO) mammalian cell expression system. It consists of 934 amino acids.
Catalog No. 50-208-4163
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.