Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethoxydiphenylsilane 98.0+%, TCI America™
SDP

Catalog No. D1731100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1731100ML 100 mL
D173125ML 25 mL
D1731500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1731100ML Supplier TCI America Supplier No. D1731100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6843-66-9
Molecular Formula C14H16O2Si
Molecular Weight (g/mol) 244.365
MDL Number MFCD00025690
InChI Key AHUXYBVKTIBBJW-UHFFFAOYSA-N
Synonym diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s
PubChem CID 81284
IUPAC Name dimethoxy(diphenyl)silane
SMILES CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC

Specifications

Color Colorless
Boiling Point 110°C
Quantity 100 mL
Formula Weight 244.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxydiphenylsilane
EINECSNumber (3)-2635
RTECSNumber VV3643332
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.