Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 2,5-Dioxahexanedioate 98.0+%, TCI America™
SDP

Catalog No. D245525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D245525G 25 g
D24555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D245525G Supplier TCI America Supplier No. D245525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88754-66-9
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00085447
InChI Key DOMLQXFMDFZAAL-UHFFFAOYSA-N
Synonym 2,5-Dioxahexanedioic Acid Dimethyl Ester
PubChem CID 263025
IUPAC Name 2-methoxycarbonyloxyethyl methyl carbonate
SMILES COC(=O)OCCOC(=O)OC

Specifications

Color Colorless
Boiling Point 220°C
Quantity 25 g
Formula Weight 178.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 2,5-Dioxahexanedioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.