Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 4-Hydroxy-2,6-pyridinedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. D49251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49251G 1 g
D49255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49251G Supplier TCI America Supplier No. D49251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19872-91-4
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00185834
InChI Key FERASHUCDLDGHK-UHFFFAOYSA-N
Synonym 4-Hydroxy-2,6-pyridinedicarboxylic Acid Dimethyl Ester
PubChem CID 592568
IUPAC Name dimethyl 4-oxo-1H-pyridine-2,6-dicarboxylate
SMILES COC(=O)C1=CC(=O)C=C(N1)C(=O)OC

Specifications

Melting Point 168°C
Color White
Quantity 1 g
Formula Weight 211.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 4-Hydroxy-2,6-pyridinedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.