Learn More
Sigma Organic Chemistry Dimethyl azelate | 500ML | 1732-10-1 | MFCD00025898 | 0.8
Supplier: Sigma Organic Chemistry 171026500ML

Dimethyl azelate, 500ML
About This Item:
Synonym(s): Dimethyl nonanedioate; Methyl azelate; Nonanedioic acid dimethyl ester
Linear Formula: CH3OCO(CH2)7COOCH3
Molecular Weight: 216.27
CAS Number: 1732-10-1
MDL Number: MFCD00025898
UNSPSC Code: 12162002
Purity: 0.8
Boiling Point: 156 C/20 mmHg (lit.)
Melting Point: 18 C
Density: 1.007 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.435 (lit.)
Storage: room temp
EINECS Number: 217-060-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.