Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Bromoterephthalate 98.0+%, TCI America™
SDP

Catalog No. D500325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D500325G 25 g
D50035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D500325G Supplier TCI America Supplier No. D500325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18643-86-2
Molecular Formula C10H9BrO4
Molecular Weight (g/mol) 273.082
MDL Number MFCD00060638
InChI Key VUMPFOPENBVFOF-UHFFFAOYSA-N
Synonym Bromoterephthalic Acid Dimethyl Ester
PubChem CID 87741
IUPAC Name dimethyl 2-bromobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)Br

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 25 g
Formula Weight 273.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dimethyl Bromoterephthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.