Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl D-(+)-Malate 98.0+%, TCI America™
SDP

Catalog No. M134725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M134725G 25 g
M13475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M134725G Supplier TCI America Supplier No. M134725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70681-41-3
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00066216
InChI Key YSEKNCXYRGKTBJ-UHFFFAOYNA-N
Synonym D-(+)-Apple Acid Dimethyl Ester, D-(+)-Malic Acid Dimethyl Ester
PubChem CID 11062697
IUPAC Name 1,4-dimethyl 2-hydroxybutanedioate
SMILES COC(=O)CC(O)C(=O)OC

Specifications

Color Colorless
Quantity 25 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl D-(+)-Malate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.