Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Decahydro-2,6-naphthalenedicarboxylate (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. D374525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D374525G 25 g
D37455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D374525G Supplier TCI America Supplier No. D374525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3068-02-8
Molecular Formula C14H22O4
Molecular Weight (g/mol) 254.33
InChI Key XYHMJVZAEWACCS-UHFFFAOYNA-N
Synonym Decahydro-2,6-naphthalenedicarboxylic Acid Dimethyl Ester
PubChem CID 11357248
IUPAC Name dimethyl 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene-2,6-dicarboxylate
SMILES COC(=O)C1CCC2CC(CCC2C1)C(=O)OC

Specifications

Color White
Boiling Point 140°C
Quantity 25 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Decahydro-2,6-naphthalenedicarboxylate (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.