Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl diselenide

Catalog No. 3285021G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-850-21G 1 g
32-850-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-850-21G Supplier Sigma Aldrich Supplier No. 3285021G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dimethyl diselenide is a volatile selenium species and its determination in biological samples by HPLC-inductively coupled argon plasma mass spectrometry was reported
  • Selenium alkylation with dimethyl diselenide was reported by solid-phase microextraction gas chromatography-mass spectrometry.dimethyl diselenide is the most powerful reagent used to identify distonic radical cations

Specifications

CAS 7101-31-7
Density 1.987 g/mL (at 25°C (literature))
Boiling Point 155°C to 157°C (lit.)
Molecular Formula C2H6Se2
Refractive Index n20/D 1.6388 (literature)
Linear Formula (CH3)2 Se2
MDL Number MFCD00008254
Quantity 1 g
Synonym Methyl diselenide
Molecular Weight (g/mol) 187.99
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.