Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Hexafluoroglutarate 96.0+%, TCI America™
SDP

Catalog No. D35891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35891G 1 g
D35895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35891G Supplier TCI America Supplier No. D35891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1513-62-8
Molecular Formula C7H6F6O4
Molecular Weight (g/mol) 268.111
MDL Number MFCD00155851
InChI Key PJZVBCPDYSZAJU-UHFFFAOYSA-N
Synonym Dimethyl Perfluoroglutarate, Dimethyl Hexafluoropentanedioate, Hexafluoroglutaric Acid Dimethyl Ester, Perfluoroglutaric Acid Dimethyl Ester, Hexafluoropentanedioic Acid Dimethyl Ester
PubChem CID 123116
IUPAC Name dimethyl 2,2,3,3,4,4-hexafluoropentanedioate
SMILES COC(=O)C(C(C(C(=O)OC)(F)F)(F)F)(F)F

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 100°C
Quantity 1 g
Formula Weight 268.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Hexafluoroglutarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.