Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Iminodiacetate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D538525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D538525G 25 g
D53855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D538525G Supplier TCI America Supplier No. D538525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39987-25-2
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.62
MDL Number MFCD00216724
InChI Key IIWYYIACSUPJCN-UHFFFAOYSA-N
Synonym dimethyl iminodiacetate hydrochloride, dimethyl 2,2'-azanediyldiacetate hydrochloride, methyl 2-2-methoxy-2-oxoethyl amino acetate hydrochloride, dimethyl iminodiacetic acid hydrochloride, iminodiacetic acid dimethyl ester hydrochloride, glycine, n-2-methoxy-2-oxoethyl-, methyl ester, hydrochloride, methyl 2-methoxycarbonyl methyl amino acetate, chloride, zlchem 797, acmc-1aeyr, iminodiacetic dimethyl ester hydrochloride
PubChem CID 2727727
IUPAC Name hydrogen methyl 2-[(2-methoxy-2-oxoethyl)amino]acetate chloride
SMILES [H+].[Cl-].COC(=O)CNCC(=O)OC

Specifications

Melting Point 169°C
Color White
Quantity 25 g
Formula Weight 197.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Iminodiacetate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.