Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl isophthalate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013138 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3138 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3138 Supplier MilliporeSigma Supelco Supplier No. 44291100MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 1459-93-4
Vapor Pressure 93.7 mmHg (208°C)
Vapor Density 6.8 (vs. air)
Flash Point 173°C
Physical Form Neat
Linear Formula C6H4-1,3-(CO2 CH3)2
MDL Number MFCD00008433
Melting Point 64°C to 68°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 194.18
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.