Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl isophthalate, 99%

Catalog No. AC186510025
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186510025 2.5 kg Plastic bottle
AC186510250 25 g Plastic bottle
AC186515000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC186510025 Supplier Thermo Scientific Chemicals Supplier No. 186510025
Only null left
Add to Cart

CAS: 1459-93-4 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1459-93-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008433
InChI Key VNGOYPQMJFJDLV-UHFFFAOYSA-N
Synonym dimethyl isophthalate, dimethyl m-phthalate, isophthalic acid dimethyl ester, methyl isophthalate, 1,3-benzenedicarboxylic acid, dimethyl ester, morflex 1129, dimethyl 1,3-benzenedicarboxylate, dimethylisophthalate, methyl 3-carbomethoxy benzoate, isophthalic acid, dimethyl ester
PubChem CID 15088
IUPAC Name dimethyl benzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC=C1)C(=O)OC

Specifications

Melting Point >66.0°C
Color White
Boiling Point 124°C (16.0 hPa)
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4(CO2CH3)2
Quantity 2.5 kg
Solubility Information Solubility in water: insoluble
Viscosity 2.14 mPa.s (99°C)
Formula Weight 194.19
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Dimethyl isophthalate, 99%
Show More Show Less
EINECSNumber 215-951-9
RTECSNumber NT2540000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.