Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Isophthalate 99.0+%, TCI America™
SDP

Catalog No. I015725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I015725G 25 g
I0157500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I015725G Supplier TCI America Supplier No. I015725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1459-93-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008433
InChI Key VNGOYPQMJFJDLV-UHFFFAOYSA-N
Synonym dimethyl isophthalate, dimethyl m-phthalate, isophthalic acid dimethyl ester, methyl isophthalate, 1,3-benzenedicarboxylic acid, dimethyl ester, morflex 1129, dimethyl 1,3-benzenedicarboxylate, dimethylisophthalate, methyl 3-carbomethoxy benzoate, isophthalic acid, dimethyl ester
PubChem CID 15088
IUPAC Name 1,3-dimethyl benzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC=C1)C(=O)OC

Specifications

Melting Point 69°C
Color White
Boiling Point 124°C
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Isophthalate
EINECSNumber (3)-1318
RTECSNumber NT2540000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.