Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals

Catalog No. AAA1426822
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1426822 100 g
AAA1426809 10 g
AAA1426836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1426822 Supplier Thermo Scientific Chemicals Supplier No. A1426822

missing translation for 'validMainframeMsg'

Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

Solubility
Miscible with water, ethanol, ether, alcohol, benzene, acetone and carbon tetrachloride. Immiscible with heavy mineral oil.

Notes
Incompatible materials strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 756-79-6
Molecular Formula C3H9O3P
Molecular Weight (g/mol) 124.08
MDL Number MFCD00008349
InChI Key VONWDASPFIQPDY-UHFFFAOYSA-N
Synonym dimethyl methylphosphonate, dimethylmethylphosphonate, pyrol dmmp, dmmp, dimethyl methanephosphonate, fyrol dmmp, phosphonic acid, methyl-, dimethyl ester, methanephosphonic acid dimethyl ester, dimethoxymethylphosphine oxide, methylphosphonic acid dimethyl ester
PubChem CID 12958
IUPAC Name [methoxy(methyl)phosphoryl]oxymethane
SMILES COP(C)(=O)OC

Specifications

Density 1.16
Boiling Point 180°C to 181°C
Flash Point 68°C (154°F)
Refractive Index 1.413
Quantity 100 g
Beilstein 878263
Merck Index 14,3391
Solubility Information Miscible with water,ethanol,ether,alcohol,benzene,acetone and carbon tetrachloride. Immiscible with heavy mineral oil.
Formula Weight 124.08
Percent Purity 97%
Chemical Name or Material Dimethyl methylphosphonate
Show More Show Less
Hazard Category H227-H319-H332-H335-H340-H360-H373
Hazard Statement GHS H Statement
H340-H319-H227
May cause genetic defects.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
EINECSNumber 212-052-3
RTECSNumber SZ9120000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.