Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals

Catalog No. AC116420250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116420250 25 g Glass bottle
AC116420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC116420250 Supplier Thermo Scientific Chemicals Supplier No. 116420250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 756-79-6
Molecular Formula C3H9O3P
Molecular Weight (g/mol) 124.08
MDL Number MFCD00008349
InChI Key VONWDASPFIQPDY-UHFFFAOYSA-N
Synonym dimethyl methylphosphonate, dimethylmethylphosphonate, pyrol dmmp, dmmp, dimethyl methanephosphonate, fyrol dmmp, phosphonic acid, methyl-, dimethyl ester, methanephosphonic acid dimethyl ester, dimethoxymethylphosphine oxide, methylphosphonic acid dimethyl ester
PubChem CID 12958
IUPAC Name [methoxy(methyl)phosphoryl]oxymethane
SMILES COP(C)(=O)OC

Specifications

Color Colorless
Density 1.1400g/mL
Boiling Point 181.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.413
Linear Formula (CH3O)2P(O)CH3
Quantity 25 g
Beilstein 04,I,572
Fieser 03,117; 11,203
Merck Index 15,3435
Specific Gravity 1.14
Formula Weight 124.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethyl methylphosphonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May cause genetic defects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 212-052-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.